Assay Details
Gene Symbol: porin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038810.2 | NP_001033899.1 | 2 - 3 | 1648 | 63 | |
NM_001259032.1 | NP_001245961.1 | 2 - 3 | 350 | 63 | ||
NM_001273436.1 | NP_001260365.1 | 2 - 3 | 410 | 63 | ||
NM_057465.4 | NP_476813.1 | 2 - 3 | 410 | 63 | ||
NM_134283.2 | NP_599110.1 | 2 - 3 | 439 | 63 | ||
GenBank mRNA | AY070509.1 | - | 1 - 2 | 2260 | 63 | |
BT024247.1 | - | 2 - 3 | 373 | 63 | ||
U70314.1 | - | 2 - 3 | 376 | 63 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
