Assay Details
| Gene Symbol: porin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038810.2 | NP_001033899.1 | 3 - 4 | 1879 | 69 | |
| NM_001259032.1 | NP_001245961.1 | 3 - 4 | 581 | 69 | ||
| NM_001273436.1 | NP_001260365.1 | 3 - 4 | 641 | 69 | ||
| NM_057465.4 | NP_476813.1 | 3 - 4 | 641 | 69 | ||
| NM_134283.2 | NP_599110.1 | 3 - 4 | 670 | 69 | ||
| GenBank mRNA | AY070509.1 | - | 2 - 3 | 2491 | 69 | |
| BT024247.1 | - | 3 - 4 | 604 | 69 | ||
| U70314.1 | - | 3 - 4 | 607 | 69 | ||
More Information
Set Membership: |
|

porin
Genomic Map