Assay Details
| Gene Symbol: Klp61F | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057470.5 | NP_476818.1 | 2 - 3 | 653 | 64 | |
| GenBank mRNA | AY069442.1 | - | 2 - 3 | 648 | 64 | |
More Information
Set Membership: |
|

Klp61F
Genomic Map