Assay Details
Gene Symbol: AP-2alpha | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057471.6 | NP_476819.2 | 2 - 3 | 598 | 78 | |
GenBank mRNA | BT032839.1 | - | 2 - 3 | 579 | 78 | |
Y11104.1 | - | 467 | 78 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
