Assay Details
| Gene Symbol: bib | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273384.2 | NP_001260313.1 | 1 - 2 | 498 | 62 | |
| NM_057489.5 | NP_476837.1 | 1 - 2 | 498 | 62 | ||
| GenBank mRNA | AY051981.1 | - | 1 - 2 | 478 | 62 | |
More Information
Set Membership: |
|

bib
Genomic Map