Assay Details
| Gene Symbol: bib | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273384.2 | NP_001260313.1 | 4 - 5 | 1000 | 69 | |
| NM_057489.5 | NP_476837.1 | 4 - 5 | 1000 | 69 | ||
| GenBank mRNA | AY051981.1 | - | 4 - 5 | 980 | 69 | |
More Information
Set Membership: |
|

bib
Genomic Map