Assay Details
Gene Symbol: grh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057496.5 | NP_476844.2 | 3 - 4 | 2494 | 75 | |
NM_057497.4 | NP_476845.2 | 3 - 4 | 3115 | 75 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
