Assay Details
| Gene Symbol: inaC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057515.3 | NP_476863.1 | 8 - 9 | 1727 | 75 | |
| GenBank mRNA | BT004495.1 | - | 8 - 9 | 1685 | 75 | |
| J04845.1 | - | 8 - 9 | 1679 | 75 | ||
More Information
Set Membership: |
|

inaC
Genomic Map