Assay Details
Gene Symbol: Su(H) | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299019.1 | NP_001285948.1 | 1 - 2 | 834 | 69 | |
NM_057520.4 | NP_476868.1 | 1 - 2 | 834 | 69 | ||
GenBank mRNA | AY069091.1 | - | 1 - 2 | 815 | 69 | |
M94383.1 | - | 1 - 2 | 812 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
