Assay Details
| Gene Symbol: msl-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299125.1 | NP_001286054.1 | 1 - 2 | 1609 | 63 | |
| NM_057548.4 | NP_476896.1 | 1 - 2 | 1609 | 63 | ||
| GenBank mRNA | BT023828.1 | - | 1 - 2 | 1604 | 63 | |
More Information
Set Membership: |
|

msl-1
Genomic Map