Assay Details
Gene Symbol: Sod2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299574.1 | NP_001286503.1 | 1 - 2 | 163 | 64 | |
NM_057577.4 | NP_476925.1 | 1 - 2 | 163 | 64 | ||
GenBank mRNA | BT004505.1 | - | 1 - 2 | 159 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
