Assay Details
| Gene Symbol: RpS13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038796.3 | NP_001033885.1 | 2 - 3 | 188 | 73 | |
| NM_001298818.1 | NP_001285747.1 | 2 - 3 | 254 | 73 | ||
| NM_057590.5 | NP_476938.1 | 2 - 3 | 201 | 73 | ||
| GenBank mRNA | AY058536.1 | - | 2 - 3 | 155 | 73 | |
| KX532032.1 | - | 2 - 3 | 155 | 73 | ||
| X91854.1 | - | 172 | 73 | |||
More Information
Set Membership: |
|

RpS13
Genomic Map