Assay Details
| Gene Symbol: Ror | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057614.3 | NP_476962.1 | 3 - 4 | 961 | 112 | |
| GenBank mRNA | BT125080.1 | - | 3 - 4 | 935 | 112 | |
| L20297.1 | - | 3 - 4 | 966 | 112 | ||
More Information
Set Membership: |
|

Ror
Genomic Map