Assay Details
Gene Symbol: Ror | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057614.3 | NP_476962.1 | 6 - 7 | 1839 | 104 | |
GenBank mRNA | BT125080.1 | - | 6 - 7 | 1813 | 104 | |
L20297.1 | - | 6 - 7 | 1844 | 104 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
