Assay Details
Gene Symbol: Sdc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001369961.1 | NP_001356968.1 | 4 - 5 | 671 | 61 | |
NM_057617.5 | NP_476965.1 | 4 - 5 | 671 | 61 | ||
GenBank mRNA | AY069377.1 | - | 4 - 5 | 664 | 61 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
