Assay Details
Gene Symbol: pelo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057634.5 | NP_476982.1 | 1 - 2 | 281 | 81 | |
GenBank mRNA | AY058644.1 | - | 1 - 2 | 298 | 81 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
