Assay Details
Gene Symbol: poe | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273293.1 | NP_001260222.1 | 11 - 12 | 14345 | 69 | |
NM_057638.4 | NP_476986.1 | 11 - 12 | 14345 | 69 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
