Assay Details
| Gene Symbol: poe | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273293.1 | NP_001260222.1 | 8 - 9 | 10267 | 65 | |
| NM_057638.4 | NP_476986.1 | 8 - 9 | 10267 | 65 | ||
More Information
Set Membership: |
|

poe
Genomic Map