Assay Details
| Gene Symbol: babo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169621.2 | NP_001163092.1 | 1 - 2 | 692 | 80 | |
| NM_057651.4 | NP_476999.1 | 1 - 2 | 692 | 80 | ||
| NM_057652.4 | NP_477000.1 | 1 - 2 | 692 | 80 | ||
| GenBank mRNA | BT009976.1 | - | 1 - 2 | 690 | 80 | |
More Information
Set Membership: |
|

babo
Genomic Map