Assay Details
Gene Symbol: babo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169621.2 | NP_001163092.1 | 2 - 3 | 1009 | 63 | |
NM_057651.4 | NP_476999.1 | 2 - 3 | 1009 | 63 | ||
NM_057652.4 | NP_477000.1 | 2 - 3 | 1009 | 63 | ||
GenBank mRNA | BT009976.1 | - | 2 - 3 | 1007 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
