Assay Details
Gene Symbol: babo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169621.2 | NP_001163092.1 | 7 - 8 | 2239 | 59 | |
NM_057651.4 | NP_476999.1 | 7 - 8 | 2320 | 59 | ||
NM_057652.4 | NP_477000.1 | 7 - 8 | 2257 | 59 | ||
GenBank mRNA | BT009976.1 | - | 7 - 8 | 2237 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
