Assay Details
Gene Symbol: babo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057652.4 | NP_477000.1 | 3 - 4 | 1126 | 77 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
