Assay Details
Gene Symbol: Mad | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273063.1 | NP_001259992.1 | 3 - 4 | 1489 | 71 | |
NM_057669.3 | NP_477017.1 | 3 - 4 | 1314 | 71 | ||
GenBank mRNA | BT004845.1 | - | 3 - 4 | 1313 | 71 | |
U10328.1 | - | 3 - 4 | 1304 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
