Assay Details
Gene Symbol: pav | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057677.4 | NP_477025.1 | 2 - 3 | 909 | 76 | |
GenBank mRNA | BT053676.1 | - | 2 - 3 | 907 | 76 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
