Assay Details
Gene Symbol: pav | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057677.4 | NP_477025.1 | 4 - 5 | 1785 | 65 | |
GenBank mRNA | BT053676.1 | - | 4 - 5 | 1783 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
