Assay Details
Gene Symbol: Pen | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057693.5 | NP_477041.1 | 2 - 3 | 444 | 68 | |
GenBank mRNA | BT003258.1 | - | 2 - 3 | 354 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
