Assay Details
Gene Symbol: RpL11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299685.1 | NP_001286614.1 | 3 - 4 | 212 | 83 | |
NM_057706.5 | NP_477054.1 | 3 - 4 | 212 | 83 | ||
GenBank mRNA | AY094790.1 | - | 2 - 3 | 176 | 83 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
