Assay Details
| Gene Symbol: Bgb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274342.1 | NP_001261271.1 | 1 | 1035 | 121 | |
| NM_057717.5 | NP_477065.3 | 1 | 1035 | 121 | ||
| GenBank mRNA | AY058784.1 | - | 1 | 1038 | 121 | |
More Information
Set Membership: |
|

Bgb
Genomic Map