Assay Details
| Gene Symbol: Sr-CI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298670.1 | NP_001285599.1 | 1 - 2 | 277 | 62 | |
| NM_057754.3 | NP_477102.1 | 1 - 2 | 277 | 62 | ||
More Information
Set Membership: |
|

Sr-CI
Genomic Map