Assay Details
Gene Symbol: crn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057770.5 | NP_477118.1 | 1 - 2 | 154 | 55 | |
GenBank mRNA | AY051666.1 | - | 1 - 2 | 86 | 55 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
