Assay Details
Gene Symbol: dpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299231.1 | NP_001286160.1 | 4 - 5 | 2056 | 58 | |
NM_057837.5 | NP_477185.1 | 4 - 5 | 1788 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
