Assay Details
| Gene Symbol: vri | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144320.2 | NP_001137792.1 | 2 - 3 | 867 | 72 | |
| NM_001298694.1 | NP_001285623.1 | 2 - 3 | 916 | 72 | ||
| NM_001298695.1 | NP_001285624.1 | 2 - 3 | 867 | 72 | ||
| NM_057843.4 | NP_477191.1 | 2 - 3 | 867 | 72 | ||
| NM_164639.2 | NP_723075.1 | 2 - 3 | 455 | 72 | ||
| GenBank mRNA | BT030402.1 | - | 2 - 3 | 455 | 72 | |
| BT031200.1 | - | 2 - 3 | 796 | 72 | ||
More Information
Set Membership: |
|

vri
Genomic Map