Assay Details
| Gene Symbol: Cdk9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057878.4 | NP_477226.2 | 1 - 2 | 317 | 95 | |
| GenBank mRNA | AF027300.1 | - | 1 - 2 | 294 | 95 | |
| AY118809.1 | - | 1 - 2 | 299 | 95 | ||
More Information
Set Membership: |
|

Cdk9
Genomic Map