Assay Details
Gene Symbol: ttv | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274042.1 | NP_001260971.1 | 2 - 3 | 1875 | 63 | |
NM_001369957.1 | NP_001356963.1 | 2 - 3 | 1566 | 63 | ||
NM_057883.3 | NP_477231.1 | 2 - 3 | 1566 | 63 | ||
GenBank mRNA | AB221351.1 | - | 2 - 3 | 1212 | 63 | |
BT021403.1 | - | 2 - 3 | 1571 | 63 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
