Assay Details
Gene Symbol: ttv | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274042.1 | NP_001260971.1 | 5 - 6 | 2197 | 85 | |
NM_001274043.1 | NP_001260972.1 | 2 - 3 | 245 | 85 | ||
NM_001369957.1 | NP_001356963.1 | 5 - 6 | 1888 | 85 | ||
NM_057883.3 | NP_477231.1 | 5 - 6 | 1888 | 85 | ||
GenBank mRNA | AB221351.1 | - | 5 - 6 | 1534 | 85 | |
BT021403.1 | - | 5 - 6 | 1893 | 85 | ||
BT124899.1 | - | 2 - 3 | 245 | 85 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BT124899.1, NM_001274043.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
