Assay Details
Gene Symbol: ttv | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274042.1 | NP_001260971.1 | 7 - 8 | 2454 | 66 | |
NM_001274043.1 | NP_001260972.1 | 4 - 5 | 502 | 66 | ||
NM_001369957.1 | NP_001356963.1 | 7 - 8 | 2145 | 66 | ||
NM_057883.3 | NP_477231.1 | 7 - 8 | 2145 | 66 | ||
GenBank mRNA | AB221351.1 | - | 7 - 8 | 1791 | 66 | |
BT021403.1 | - | 7 - 8 | 2150 | 66 | ||
BT124899.1 | - | 4 - 5 | 502 | 66 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
