Assay Details
| Gene Symbol: Dredd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001297822.1 | NP_001284751.1 | 2 - 3 | 909 | 80 | |
| NM_057901.4 | NP_477249.3 | 2 - 3 | 890 | 80 | ||
More Information
Set Membership: |
|

Dredd
Genomic Map