Assay Details
| Gene Symbol: Dredd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001297822.1 | NP_001284751.1 | 3 - 4 | 1272 | 67 | |
| NM_057901.4 | NP_477249.3 | 3 - 4 | 1253 | 67 | ||
| NM_057902.4 | NP_477250.3 | 2 - 3 | 1311 | 67 | ||
| NM_057903.4 | NP_477251.3 | 3 - 4 | 1235 | 67 | ||
More Information
Set Membership: |
|

Dredd
Genomic Map