Assay Details
Gene Symbol: Dredd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001297822.1 | NP_001284751.1 | 4 - 5 | 1437 | 107 | |
NM_057901.4 | NP_477249.3 | 4 - 5 | 1418 | 107 | ||
NM_057902.4 | NP_477250.3 | 3 - 4 | 1476 | 107 | ||
NM_057903.4 | NP_477251.3 | 4 - 5 | 1400 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
