Assay Details
Gene Symbol: PheRS-m | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057935.5 | NP_477283.1 | 1 - 2 | 1290 | 79 | |
GenBank mRNA | AY058580.1 | - | 1 - 2 | 1251 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
