Assay Details
Gene Symbol: Rpn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273628.1 | NP_001260557.1 | 1 - 2 | 290 | 82 | |
NM_057952.4 | NP_477300.1 | 1 - 2 | 429 | 82 | ||
GenBank mRNA | BT044249.1 | - | 1 - 2 | 389 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
