Assay Details
Gene Symbol: ebi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057981.4 | NP_477329.1 | 2 - 3 | 1582 | 56 | |
GenBank mRNA | AF146345.1 | - | 2 - 3 | 1578 | 56 | |
BT133073.1 | - | 2 - 3 | 1547 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
