Assay Details
Gene Symbol: Abl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104153.3 | NP_001097623.1 | 6 - 7 | 2947 | 80 | |
NM_001275030.1 | NP_001261959.1 | 5 - 6 | 2893 | 80 | ||
NM_001275031.1 | NP_001261960.1 | 6 - 7 | 2947 | 80 | ||
NM_001275032.1 | NP_001261961.1 | 5 - 6 | 2893 | 80 | ||
NM_001275033.1 | NP_001261962.1 | 6 - 7 | 2947 | 80 | ||
NM_001275034.1 | NP_001261963.1 | 6 - 7 | 2947 | 80 | ||
NM_001275035.1 | NP_001261964.1 | 5 - 6 | 2893 | 80 | ||
NM_001275036.1 | NP_001261965.1 | 6 - 7 | 2947 | 80 | ||
NM_001300156.1 | NP_001287085.1 | 5 - 6 | 2893 | 80 | ||
NM_080104.3 | NP_524843.2 | 5 - 6 | 2893 | 80 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
