Assay Details
Gene Symbol: e(y)2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080107.3 | NP_524846.1 | 1 | 361 | 91 | |
GenBank mRNA | AF173295.1 | - | 1 | 336 | 91 | |
BT022196.1 | - | 1 | 307 | 91 | ||
KX531262.1 | - | 1 | 219 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
