Assay Details
Gene Symbol: ImpE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274648.1 | NP_001261577.1 | 5 - 6 | 4640 | 75 | |
NM_080110.3 | NP_524849.2 | 5 - 6 | 4584 | 75 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
