Assay Details
Gene Symbol: CanB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259263.2 | NP_001246192.1 | 3 - 4 | 590 | 81 | |
NM_080135.4 | NP_524874.2 | 3 - 4 | 590 | 81 | ||
GenBank mRNA | AY089602.1 | - | 3 - 4 | 588 | 81 | |
BT003768.1 | - | 3 - 4 | 572 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
