Assay Details
Gene Symbol: comt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272577.2 | NP_001259506.1 | 2 - 3 | 323 | 67 | |
NM_080138.4 | NP_524877.1 | 2 - 3 | 323 | 67 | ||
GenBank mRNA | BT010259.1 | - | 2 - 3 | 348 | 67 | |
U09373.1 | - | 2 - 3 | 377 | 67 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
