Assay Details
| Gene Symbol: dup | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299522.1 | NP_001286451.1 | 2 - 3 | 956 | 89 | |
| NM_080139.5 | NP_524878.1 | 2 - 3 | 1330 | 89 | ||
| GenBank mRNA | AF279146.1 | - | 2 - 3 | 952 | 89 | |
| AY118555.1 | - | 2 - 3 | 1292 | 89 | ||
More Information
Set Membership: |
|

dup
Genomic Map