Assay Details
| Gene Symbol: Tor | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273498.1 | NP_001260427.1 | 1 - 2 | 532 | 92 | |
| NM_080152.3 | NP_524891.1 | 1 - 2 | 532 | 92 | ||
More Information
Set Membership: |
|

Tor
Genomic Map