Assay Details
| Gene Symbol: Tor | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080152.3 | NP_524891.1 | 3 - 4 | 1322 | 80 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tor| Gene Symbol: Tor | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080152.3 | NP_524891.1 | 3 - 4 | 1322 | 80 | |
Set Membership: |
|